Learn More
Tri-n-butyl phosphite, 94%, Thermo Scientific Chemicals
Description
Tributyl phosphite may be employed as model lubricant additive and its surface chemistry on Fe3O4 in ultrahigh vacuum is investigated. It is used as lubricants, preservatives, heat stabilizers of intensifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tributyl phosphite may be employed as model lubricant additive and its surface chemistry on Fe3O4 in ultrahigh vacuum is investigated. It is used as lubricants, preservatives, heat stabilizers of intensifier.
Solubility
Not miscible or difficult to mix in water.
Notes
Air and moisture sensitive. Store under inert gas. Store away from oxidizing agent, air, water/ moisture.
Specifications
Melting Point | -80°C |
Density | 0.915 |
Boiling Point | 125°C to 128°C (10 mmHg) |
Flash Point | 82°C (179°F) |
Refractive Index | 1.432 |
Quantity | 100 mL |
Beilstein | 1703866 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 250.32 |
Show More |
Safety and Handling
Hazard Category | H227-H312+H332-H315-H319 |
Hazard Statement | GHS H Statement H312-H315-H319-H227 Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Combustible liquid. |
Precautionary Statement | P210-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 203-061-3 |
RTECSNumber | TH0875000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.