Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tri-n-butylborane, 1M soln. in THF

Manufacturer:  Alfa Aesar™ H32518AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH32518AE


  Check Availability
Add to cart

Specifications

Description

Applications
Tri-n-butylborane is used as a reactant for addition reactions for synthesis of phenyltrialkylborate salts, boron-based multicomponent reactions for generation of substituted aziridines, oxazolidines, and pyrrolidines, Suzuki-Miyaura cross coupling of halopurines with trialkylboranes for the synthesis of adenosine A2A receptor antagonists, multicomponent C-alkylation reactions with aromatic aldimines, one-pot alkylation-boration and electrophilic substitution.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

122-56-5
182.16
UN2924
C12H27B
MFCD00009423
Tri-n-butylborane

Specifications

1M soln. in THF
122-56-5
182.16
UN2924
100mL
C12H27B
MFCD00009423
Tri-n-butylborane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H351-H302-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit