Alfa Aesar™ Tri-n-butylborane, 1M soln. in THF

Manufacturer: Alfa Aesar™ H32518AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH32518AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 122-56-5
    Molecular Formula C12H27B
    Formula Weight 182.16
    MDL Number MFCD00009423
    UN Number UN2924
    Chemical Name or Material Tri-n-butylborane

    Specifications

    Formula Weight 182.16
    TSCA TSCA
    MDL Number MFCD00009423
    UN Number UN2924
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration 1M soln. in THF
    Quantity 500mL
    Molecular Formula C12H27B
    CAS 122-56-5
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H351-H302-H319-H335
    Highly flammable liquid and vapor.
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material Tri-n-butylborane

    Description

    Applications
    Tri-n-butylborane is used as a reactant for addition reactions for synthesis of phenyltrialkylborate salts, boron-based multicomponent reactions for generation of substituted aziridines, oxazolidines, and pyrrolidines, Suzuki-Miyaura cross coupling of halopurines with trialkylboranes for the synthesis of adenosine A2A receptor antagonists, multicomponent C-alkylation reactions with aromatic aldimines, one-pot alkylation-boration and electrophilic substitution.

    Solubility
    Not miscible or difficult to mix with water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H225-H351-H302-H319-H335
    Highly flammable liquid and vapor.
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit