Alfa Aesar™ Tri-n-butylborane, 98%

Manufacturer: Alfa Aesar™ 3353309

Quantity 10g

 View more versions of this product

Catalog No. AA3353309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 122-56-5
    Formula Weight 182.16
    MDL Number MFCD00009423
    UN Number UN2845
    Synonym Tri-n-butylboron
    Chemical Name or Material Tri-n-butylborane

    Specifications

    Formula Weight 182.16
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD00009423
    RTECS Number ED1850000
    Refractive Index 1.4285
    CAS 122-56-5
    Health Hazard 1 Danger
    Density 0.747g/mL
    UN Number UN2845
    Synonym Tri-n-butylboron
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Flash Point −20° (−4°F)
    Sensitivity Air Sensitive
    Boiling Point 170°C (222mmHg to 225mmHg)
    Melting Point −34°C
    Quantity 10g
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H250-H314-H302
    Catches fire spontaneously if exposed to air.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Chemical Name or Material Tri-n-butylborane
    Assay 98%

    GHS H Statement
    H250-H314-H302
    Catches fire spontaneously if exposed to air.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit