Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tri-n-butylborane, 98%

Manufacturer:  Alfa Aesar™ 3353306

Quantity 5g

 View more versions of this product

Catalog No. AA3353306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

122-56-5
MFCD00009423
Tri-n-butylboron
182.16
UN2845
Tri-n-butylborane

Specifications

0.747g/mL
Air Sensitive
−34°C
122-56-5
Liquid
MFCD00009423
Tri-n-butylboron
98%
−20° (−4°F)
170°C (222mmHg to 225mmHg)
5g
1.4285
182.16
UN2845
Tri-n-butylborane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H250-H314-H302
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : ED1850000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit