Alfa Aesar™ Tri-n-butylphosphine, 94%

Manufacturer: Alfa Aesar™ 1411418

Quantity 50g

 View more versions of this product

Catalog No. AA1411418

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 998-40-3
    Formula Weight 202.32
    MDL Number MFCD00009462
    UN Number UN3254
    Beilstein 1738261
    Chemical Name or Material Tri-n-butylphosphine

    Specifications

    Formula Weight 202.32
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H250-H252-H314-H318-H302-H312
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    UN Number UN3254
    RTECS Number SZ3270000
    Quantity 50g
    Refractive Index 1.4620
    Physical Form Liquid
    MDL Number MFCD00009462
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: TRIBUTYLPHOSPHANE
    Sensitivity Air & Moisture Sensitive
    Boiling Point 240°C
    TSCA TSCA
    Beilstein 1738261
    Solubility Information Insoluble in water; Miscible with ether, methanol, ethanol and benzene
    Density 0.820g/mL
    Flash Point >93° (199°F)
    Melting Point −65°C
    Odor Unpleasant
    CAS 998-40-3
    Chemical Name or Material Tri-n-butylphosphine
    Assay 94%

    Description

    • For cleavage of disulfide bonds, as a polymerization crosslinking catalyst, as an organic intermediate, and in fuels

    GHS H Statement
    H250-H252-H314-H318-H302-H312
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit