Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tri-n-butyltin acetate, 98%

Click to view available options
Quantity:
1000g
5000g
56-36-0
C14H30O2Sn
349.102
MFCD00015228
PWBHRVGYSMBMIO-UHFFFAOYSA-M
tri-n-butyltin acetate,tributyltin acetate,acetoxytributyltin,tribuyltin acetate,acetoxytributylstannane,tributylacetoxystannane,tin, acetoxytributyl,stannane, acetyloxy tributyl,stannane, acetoxytributyl,tributylstannium acetate
16682741
tributylstannyl acetate
CCCC[Sn](CCCC)(CCCC)OC(=O)C
56-36-0
C14H30O2Sn
349.102
MFCD00015228
PWBHRVGYSMBMIO-UHFFFAOYSA-M
tri-n-butyltin acetate,tributyltin acetate,acetoxytributyltin,tribuyltin acetate,acetoxytributylstannane,tributylacetoxystannane,tin, acetoxytributyl,stannane, acetyloxy tributyl,stannane, acetoxytributyl,tributylstannium acetate
16682741
tributylstannyl acetate
CCCC[Sn](CCCC)(CCCC)OC(=O)C
Melting Point 86°C to 87°C
Quantity 1000g
UN Number UN3146
Formula Weight 349.08
Percent Purity 98%
Chemical Name or Material Tri-n-butyltin acetate

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

EINECSNumber : 200-269-6

TSCA : No

Recommended Storage : Ambient temperatures