Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tri-n-butyltin benzoate, 98%

Manufacturer:  Alfa Aesar™ H55449AC

Quantity 25mL

 View more versions of this product

Catalog No. AAH55449AC

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

4342-36-3
411.15
UN2788
C6H5CO2Sn[(CH2)3CH3]3
MFCD00078009
Tri-n-butyltin benzoate

Specifications

1.193g/mL
4342-36-3
Liquid
411.15
UN2788
98%
25mL
1.5180
C6H5CO2Sn[(CH2)3CH3]3
MFCD00078009
Tri-n-butyltin benzoate

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P362-P312-P363-P321-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

RTECSNumber : WH6710000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit