Alfa Aesar™ Tri-n-butyltin benzoate, 98%

Manufacturer: Alfa Aesar™ H55449AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH55449AE

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 4342-36-3
    Linear Formula C6H5CO2Sn[(CH2)3CH3]3
    Formula Weight 411.15
    MDL Number MFCD00078009
    UN Number UN2788
    Chemical Name or Material Tri-n-butyltin benzoate

    Specifications

    Formula Weight 411.15
    Health Hazard 1 Danger
    TSCA TSCA
    Refractive Index 1.5180
    Physical Form Liquid
    MDL Number MFCD00078009
    DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Quantity 100mL
    CAS 4342-36-3
    UN Number UN2788
    Density 1.193g/mL
    Linear Formula C6H5CO2Sn[(CH2)3CH3]3
    Health Hazard 2 GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER o
    RTECS Number WH6710000
    Chemical Name or Material Tri-n-butyltin benzoate
    Assay 98%

    GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER o

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit