Alfa Aesar™ Tri-n-butyltin bromide, 95%

Manufacturer: Alfa Aesar™ 7118514

Quantity 25g

 View more versions of this product

Catalog No. AA7118514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1461-23-0
    Molecular Formula [CH3(CH2)3]3SnBr
    Formula Weight 369.96
    MDL Number MFCD00010660
    UN Number UN2788
    Synonym Bromotributyltin
    Chemical Name or Material Tri-n-butyltin bromide

    Specifications

    Formula Weight 369.96
    Health Hazard 3 GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00010660
    CAS 1461-23-0
    Flash Point >110°C (230°F)
    Density 1.338g/mL
    Refractive Index 1.5070
    RTECS Number WH6735000
    Quantity 25g
    Synonym Bromotributyltin
    Health Hazard 1 Danger
    Sensitivity Moisture Sensitive
    Physical Form Liquid
    Molecular Formula [CH3(CH2)3]3SnBr
    UN Number UN2788
    Health Hazard 2 GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Boiling Point 163°C (12mmHg)
    Chemical Name or Material Tri-n-butyltin bromide
    Assay 95%

    GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit