Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyltin chloride, 95%, tech., ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
1461-22-9
C12H27ClSn
325.48
MFCD00000521
GCTFWCDSFPMHHS-UHFFFAOYSA-M
tributyltin chloride,chlorotributyltin,tri-n-butyltin chloride,tributylchlorotin,stannane, tributylchloro,chlorotri-n-butyltin,tri-n-butylchlorotin,tributyl chloro stannane,chlorotributylstannane,tributylstannyl chloride
15096
CHEBI:79734
tributyl(chloro)stannane
CCCC[Sn](CCCC)(CCCC)Cl
1461-22-9
C12H27ClSn
325.48
MFCD00000521
GCTFWCDSFPMHHS-UHFFFAOYSA-M
tributyltin chloride,chlorotributyltin,tri-n-butyltin chloride,tributylchlorotin,stannane, tributylchloro,chlorotri-n-butyltin,tri-n-butylchlorotin,tributyl chloro stannane,chlorotributylstannane,tributylstannyl chloride
15096
CHEBI:79734
tributyl(chloro)stannane
CCCC[Sn](CCCC)(CCCC)Cl
Melting Point -9.0°C
Density 1.2000g/mL
Color Colorless to Yellow
Infrared Spectrum Authentic
Boiling Point 171.0°C to 173.0°C (25.0mmHg)
Flash Point >112°C
Quantity 100g
Specific Gravity 1.2
Refractive Index 1.4880 to 1.4940
Viscosity 7 mPa.s (20°C)
Packaging Glass bottle
Assay Percent Range 95%
Linear Formula [CH3(CH2)3]3SnCl
Merck Index 15, 9782
Solubility Information Solubility in water: 0.07g/L (20°C). Other solubilities: soluble in most common orgnaic solvents,including,benzene and toluene
Formula Weight 325.48
Physical Form Liquid
Percent Purity 95%
Grade Technical
Chemical Name or Material Tri-n-butyltin chloride, tech. 90%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 215-958-7

RTECSNumber : WH6820000

TSCA : TSCA