Alfa Aesar™ Tri-n-butyltin deuteride, 96%

Manufacturer: Alfa Aesar™ 8847403

Quantity 1g

 View more versions of this product

Catalog No. AA8847403

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 6180-99-0
    Molecular Formula C12H27DSn
    Formula Weight 292.06
    MDL Number MFCD00064886
    UN Number UN2929
    Synonym Deutero tri-n-butylstannane
    Chemical Name or Material Tri-n-butyltin deuteride

    Specifications

    Formula Weight 292.06
    Health Hazard 2 GHS H Statement
    H301-H372-H226-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Flammable liquid and vapor.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P273-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Recognizable
    Molecular Formula C12H27DSn
    CAS 6180-99-0
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
    Density 1.103g/mL
    Sensitivity Air & Moisture Sensitive
    Refractive Index 1.4720
    Physical Form Liquid
    UN Number UN2929
    Health Hazard 1 Danger
    Flash Point 40°C (104°F)
    MDL Number MFCD00064886
    Synonym Deutero tri-n-butylstannane
    Boiling Point 65°C (0.6mmHg)
    Color Colorless
    Quantity 1g
    Chemical Name or Material Tri-n-butyltin deuteride
    Assay 96%

    GHS H Statement
    H301-H372-H226-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Flammable liquid and vapor.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P273-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit