Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyltin methoxide, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
1067-52-3
C13H30OSn
321.07
MFCD00009419
KJGLZJQPMKQFIK-UHFFFAOYSA-N
tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl
16683411
tributyl(methoxy)stannane
CCCC[Sn](CCCC)(CCCC)OC
1067-52-3
C13H30OSn
321.07
MFCD00009419
KJGLZJQPMKQFIK-UHFFFAOYSA-N
tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl
16683411
tributyl(methoxy)stannane
CCCC[Sn](CCCC)(CCCC)OC
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 97°C to 97.5°C
Specific Gravity 1.11
Flash Point 98°C
Refractive Index 1.4726 to 1.4746
Density 1.1100g/mL
Quantity 25g
Color Colorless
Elemental Analysis 96.0% min.
Assay Percent Range 97%
Linear Formula [CH3(CH2)3]3SnOCH3
Fieser 16,351
Merck Index 15, 9784
Formula Weight 321.07
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin methoxide
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear p

GHS Signal Word: Danger

EINECSNumber : 213-933-5