Alfa Aesar™ Tri-n-butyltin trifluoromethanesulfonate, tech. 90%

Manufacturer: Alfa Aesar™ H55192MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55192MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 68725-14-4
    Linear Formula CF3SO3Sn[(CH2)3CH3]3
    Formula Weight 439.1
    MDL Number MFCD00043324
    UN Number UN2923
    Beilstein 5818090
    Synonym Tri-n-butyltin triflate
    Chemical Name or Material Tri-n-butyltin trifluoromethanesulfonate
    Grade Technical Grade

    Specifications

    Formula Weight 439.1
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00043324
    UN Number UN2923
    Synonym Tri-n-butyltin triflate
    Health Hazard 2 GHS H Statement
    H301-H372-H314-H318-H312
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful in contact with skin.
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Physical Form Solid
    Linear Formula CF3SO3Sn[(CH2)3CH3]3
    CAS 68725-14-4
    Beilstein 5818090
    Health Hazard 1 Danger
    Color White to pale brown
    Sensitivity Moisture Sensitive
    Quantity 250mg
    Chemical Name or Material Tri-n-butyltin trifluoromethanesulfonate
    Assay 90%
    Grade Technical Grade

    GHS H Statement
    H301-H372-H314-H318-H312
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful in contact with skin.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit