Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tri-n-hexylamine, 97%

Click to view available options
Quantity:
50g
250g
102-86-3
C18H39N
269.517
MFCD00009523
DIAIBWNEUYXDNL-UHFFFAOYSA-N
tri-n-hexylamine,trihexylamine,1-hexanamine, n,n-dihexyl,c6-c12 trialkylamine,amines, tri-c6-12-alkyl,amine,trihexyl,1-hexanamine,n-dihexyl,1-hexanamine,n,n-dihexyl
66022
N,N-dihexylhexan-1-amine
CCCCCCN(CCCCCC)CCCCCC
102-86-3
C18H39N
269.517
MFCD00009523
DIAIBWNEUYXDNL-UHFFFAOYSA-N
tri-n-hexylamine,trihexylamine,1-hexanamine, n,n-dihexyl,c6-c12 trialkylamine,amines, tri-c6-12-alkyl,amine,trihexyl,1-hexanamine,n-dihexyl,1-hexanamine,n,n-dihexyl
66022
N,N-dihexylhexan-1-amine
CCCCCCN(CCCCCC)CCCCCC
Applications
Tri-n-hexylamine is used as an extractant during isolation and purification of succinic acid from an escherichia coli fermentation broth in pharmaceutical industry. It is also used as an internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. Further, it is involved in the synthesis of zinc oxide nanorods. In addition to this, it is used in the preparation of infrared luminescence nanohybrid films.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, acids, acid chloridres, acid anhydrides and chloroformates.
Flash Point 144°C (291°F)
Melting Point <-75°C
Boiling Point 263°C to 265°C
Quantity 50g
UN Number UN3082
Beilstein 1755828
Formula Weight 269.52
Percent Purity 97%
Chemical Name or Material Tri-n-hexylamine

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA