Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-octylamine, 97%, Thermo Scientific Chemicals

Catalog Number AC140281000
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
1116-76-3
C24H51N
353.66
XTAZYLNFDRKIHJ-UHFFFAOYSA-N
trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
14227
N,N-dioctyloctan-1-amine
CCCCCCCCN(CCCCCCCC)CCCCCCCC
1116-76-3
C24H51N
353.66
XTAZYLNFDRKIHJ-UHFFFAOYSA-N
trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
14227
N,N-dioctyloctan-1-amine
CCCCCCCCN(CCCCCCCC)CCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -34.0°C
Color White
Density 0.8090g/mL
Boiling Point 365.0°C to 367.0°C
Flash Point 163°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4480 to 1.4500
Linear Formula [CH3(CH2)7]3N
Quantity 100 mL
Beilstein 04,196
Specific Gravity 0.809
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol
Formula Weight 353.66
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Tri-n-octylamine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the envir
EINECSNumber 214-242-1

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit