Learn More
Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals
Description
Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.
Solubility
Soluble in chloroform, toluene and hexane. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 52°C to 57°C |
Density | 0.88 |
Boiling Point | 212°C to 213°C (0.3 mmHg) |
Flash Point | 252°C (486°F) |
Odor | Odorless |
Quantity | 500 g |
Beilstein | 1796648 |
Solubility Information | Soluble in chloroform,toluene and hexane. Insoluble in water. |
Formula Weight | 386.65 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H315-H317-H361 |
Hazard Statement | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
Precautionary Statement | P201-P202-P261-P264b-P272-P280-P281-P302+P352-P305+P351+P338-P308+P313-P333+P313-P362-P501c |
EINECSNumber | 201-121-3 |
RTECSNumber | SZ1662500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.