Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-octylphosphine oxide, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
78-50-2
C24H51OP
386.64
MFCD00002083
ZMBHCYHQLYEYDV-UHFFFAOYSA-N
trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg
65577
1-dioctylphosphoryloctane
CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC
78-50-2
C24H51OP
386.64
MFCD00002083
ZMBHCYHQLYEYDV-UHFFFAOYSA-N
trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg
65577
1-dioctylphosphoryloctane
CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC
Boiling Point 201.0°C to 202.0°C (2.0mmHg)
Quantity 25g
Packaging Glass bottle
Color White to Yellow
Flash Point 230°C
Melting Point 50.0°C to 55.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (including isomer) (GC)
Linear Formula [CH3(CH2)7]3P(O)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in cyclohexane,methanol
Formula Weight 386.64
Physical Form Crystalline Powder, Crystals or Flakes
Percent Purity 99%
Chemical Name or Material Tri-n-octylphosphine oxide
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Suspected of damaging fertility or the unborn child.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protecti

GHS Signal Word: Warning

EINECSNumber : 201-121-3