Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tri-n-octylphosphine oxide, 99%, ACROS Organics™
Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
Specifications
Boiling Point | 201.0°C to 202.0°C (2.0mmHg) |
Melting Point | 50.0°C to 55.0°C |
Quantity | 100g |
Flash Point | 230°C |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Color | White to Yellow |
Assay Percent Range | 98.5% min. (including isomer) (GC) |
Linear Formula | [CH3(CH2)7]3P(O) |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in cyclohexane,methanol |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Suspected of damaging fertility or the unborn child.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protecti
GHS Signal Word: Warning
EINECSNumber : 201-121-3