Alfa Aesar™ Tri-n-propyl borate, 98%

Manufacturer: Alfa Aesar™ 22974A1

Quantity 1kg

 View more versions of this product

Catalog No. AA22974A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 688-71-1
    Linear Formula (C3H7O)3B
    Formula Weight 188.08
    MDL Number MFCD00009366
    UN Number UN3272
    Synonym n-Propyl borate
    Chemical Name or Material Tri-n-propyl borate

    Specifications

    Formula Weight 188.08
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3272
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number ED5785000
    Refractive Index 1.3950
    Linear Formula (C3H7O)3B
    CAS 688-71-1
    Synonym n-Propyl borate
    Solubility Information Soluble in organic solvents; Decomposes in water
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Flash Point 32° (90°F)
    Physical Form Liquid
    MDL Number MFCD00009366
    Density 0.861g/mL
    Boiling Point 175° to 177°C
    TSCA TSCA
    Health Hazard 1 Warning
    Sensitivity Moisture Sensitive
    Color Colorless
    Quantity 1kg
    Chemical Name or Material Tri-n-propyl borate
    Assay 98%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit