Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tri-n-propylphosphine, 98%

Manufacturer:  Alfa Aesar™ H27838-14

Quantity 25g

 View more versions of this product

Catalog No. AAH2783814

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

2234-97-1
160.24
UN2845
(CH3CH2CH2)3P
MFCD00015217
Tri-n-propylphosphine

Specifications

62°C (144°F)
25g
(CH3CH2CH2)3P
MFCD00015217
Tri-n-propylphosphine
72° to 74°C (12mmHg)
2234-97-1
160.24
UN2845
98%

Safety and Handling

GHS H Statement
H250-H252-H314-H318-H227
Catches fire spontaneously if exposed to air.
Self-heating in large quantities; may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit