Alfa Aesar™ Tri-n-propylphosphine, 98%

Manufacturer: Alfa Aesar™ H27838-22

Quantity 100g

 View more versions of this product

Catalog No. AAH2783822

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2234-97-1
    Molecular Formula (CH3CH2CH2)3P
    Formula Weight 160.24
    MDL Number MFCD00015217
    UN Number UN2845
    Chemical Name or Material Tri-n-propylphosphine

    Specifications

    Formula Weight 160.24
    MDL Number MFCD00015217
    CAS 2234-97-1
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Boiling Point 72° to 74°C (12mmHg)
    Quantity 100g
    Molecular Formula (CH3CH2CH2)3P
    UN Number UN2845
    Health Hazard 2 GHS H Statement
    H250-H252-H314-H318-H227
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities; may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 62°C (144°F)
    Chemical Name or Material Tri-n-propylphosphine
    Assay 98%

    GHS H Statement
    H250-H252-H314-H318-H227
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities; may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit