Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-propylsilane, 99%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
998-29-8
C9H22Si
158.36
MFCD00009358
MMYRBBZVCDXGHG-UHFFFAOYSA-N
tripropylsilane,tri-n-propylsilane,silane, tripropyl,silicon tri-n-propyl,tripropyl silane,tri-n-propyl silane,zhovawfvvbwegq-uhfffaoysa-n
6327709
tripropylsilicon
CCC[Si](CCC)CCC
998-29-8
C9H22Si
158.36
MFCD00009358
MMYRBBZVCDXGHG-UHFFFAOYSA-N
tripropylsilane,tri-n-propylsilane,silane, tripropyl,silicon tri-n-propyl,tripropyl silane,tri-n-propyl silane,zhovawfvvbwegq-uhfffaoysa-n
6327709
tripropylsilicon
CCC[Si](CCC)CCC
Reduction reagent
Boiling Point 173°C
Density 0.7580g/mL
Refractive Index 1.4270 to 1.429
Flash Point 43°C
Specific Gravity 0.758
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Color Colorless
Assay Percent Range 98.5% min. (GC)
Linear Formula HSi(CH2CH2CH3)3
Solubility Information Solubility in water: insoluble
Formula Weight 158.36
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Tri-n-propylsilane
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-649-1