Alfa Aesar™ Tri-n-propyltin chloride

Manufacturer: Alfa Aesar™ 7112206

Quantity 5g

 View more versions of this product

Catalog No. AA7112206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2279-76-7
    Molecular Formula (CH3CH2CH2)3SnCl
    Formula Weight 283.41
    MDL Number MFCD00013610
    UN Number UN2788
    Chemical Name or Material Tri-n-propyltin chloride

    Specifications

    Formula Weight 283.41
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Health Hazard 1 Danger
    Refractive Index 1.4910
    MDL Number MFCD00013610
    UN Number UN2788
    Health Hazard 2 GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Density 1.2678g/mL at 28°C
    Sensitivity Moisture Sensitive
    Boiling Point 123°C (13mmHg)
    Melting Point −23.5°C
    Physical Form Liquid
    Molecular Formula (CH3CH2CH2)3SnCl
    CAS 2279-76-7
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Chemical Name or Material Tri-n-propyltin chloride

    GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit