Alfa Aesar™ Tri(o-tolyl) phosphite, typically C 71%, H 6%

Manufacturer: Alfa Aesar™ 1025814

Quantity 25g

 View more versions of this product

Catalog No. AA1025814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2622-08-4
    Formula Weight 352.36
    MDL Number MFCD00014911
    Synonym Tri-o-cresyl phosphite; Tris(2-tolyl)phosphite
    Chemical Name or Material Tri(o-tolyl) phosphite

    Specifications

    Formula Weight 352.36
    MDL Number MFCD00014911
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Physical Form Liquid
    CAS 2622-08-4
    Synonym Tri-o-cresyl phosphite; Tris(2-tolyl)phosphite
    Health Hazard 1 Warning
    Flash Point >110° (230°F)
    Sensitivity Air Sensitive
    Boiling Point 200° to 202°C (2.4mmHg to 2.6mmHg)
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    RTECS Number TH1050000
    Color Colorless
    Concentration Typically C 71%, H 6%
    Chemical Name or Material Tri(o-tolyl) phosphite

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit