Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-tert-butylphosphonium tetrafluoroborate, 97+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
131274-22-1
C12H28P·BF4
290.13
MFCD04039975
YTJUCJAUJCXFTN-UHFFFAOYSA-O
tri-tert-butylphosphonium tetrafluoroborate,t-bu 3ph bf4,tri-t-butylphosphonium tetrafluoroborate,fluoroboric acid tri-tert-butylphosphine adduct,tri-tert-butylphosphanium tetrafluoroborate,tbu3p hbf4,ttbp hbf4,t-bu3p hbf4,tbu 3p hbf4,acmc-20a4f9
2734635
tritert-butylphosphanium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)(C)[PH+](C(C)(C)C)C(C)(C)C
131274-22-1
C12H28P·BF4
290.13
MFCD04039975
YTJUCJAUJCXFTN-UHFFFAOYSA-O
tri-tert-butylphosphonium tetrafluoroborate,t-bu 3ph bf4,tri-t-butylphosphonium tetrafluoroborate,fluoroboric acid tri-tert-butylphosphine adduct,tri-tert-butylphosphanium tetrafluoroborate,tbu3p hbf4,ttbp hbf4,t-bu3p hbf4,tbu 3p hbf4,acmc-20a4f9
2734635
tritert-butylphosphanium;tetrafluoroborate
[B-](F)(F)(F)F.CC(C)(C)[PH+](C(C)(C)C)C(C)(C)C
Melting Point 261.0°C
NMR 97.0% min.
Color White
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97+%
Formula Weight 290.13
Physical Form Crystals and Chunks
Percent Purity 97+%
Chemical Name or Material Tri-tert-butylphosphonium tetrafluoroborate
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.