Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triacetoxy(ethyl)silane, 96%

Click to view available options
Quantity:
100g
500g
17689-77-9
C8H14O6Si
234.279
MFCD00053703
KXJLGCBCRCSXQF-UHFFFAOYSA-N
ethyltriacetoxysilane,triacetoxyethylsilane,triacetoxy ethyl silane,triacetoxy ethylsilane,ethylsilanetriyl triacetate,silanetriol, ethyl-, triacetate,silanetriol, 1-ethyl-, 1,1,1-triacetate,dsstox_cid_7792,dsstox_rid_78567,dsstox_gsid_27792
87244
[diacetyloxy(ethyl)silyl] acetate
CC[Si](OC(=O)C)(OC(=O)C)OC(=O)C
17689-77-9
C8H14O6Si
234.279
MFCD00053703
KXJLGCBCRCSXQF-UHFFFAOYSA-N
ethyltriacetoxysilane,triacetoxyethylsilane,triacetoxy ethyl silane,triacetoxy ethylsilane,ethylsilanetriyl triacetate,silanetriol, ethyl-, triacetate,silanetriol, 1-ethyl-, 1,1,1-triacetate,dsstox_cid_7792,dsstox_rid_78567,dsstox_gsid_27792
87244
[diacetyloxy(ethyl)silyl] acetate
CC[Si](OC(=O)C)(OC(=O)C)OC(=O)C
Applications
Triacetoxy(ethyl)silane is used in sealants, the substance is used as a cross-linking agent and adhesion promoter.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
UN Number UN1760
Formula Weight 234.28
Percent Purity 96%
Boiling Point 97°C to 99°C (4mmHg)
Melting Point 7°C to 9°C
Quantity 100g
Flash Point 106°C (224°F)
Chemical Name or Material Triacetoxy(ethyl)silane

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.

TSCA : TSCA