Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triacetoxy(methyl)silane, 96%

Manufacturer:  Alfa Aesar™ B2397409

Quantity 10g

 View more versions of this product

Catalog No. AAB2397409


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

4253-34-3
220.25
UN1759
Methyltriacetoxysilane
C7H12O6Si
MFCD00008694
1788668
Triacetoxy(methyl)silane

Specifications

4253-34-3
220.25
UN1759
Methyltriacetoxysilane
40° to 45°C
85°C (185°F)
96%
C7H12O6Si
MFCD00008694
1788668
87° to 88°C (3mmHg)
10g
Triacetoxy(methyl)silane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

RTECSNumber : VV4500000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit