Alfa Aesar™ Triacetoxy(vinyl)silane, 95%

Manufacturer: Alfa Aesar™ B242120B

Quantity 1000g

 View more versions of this product

Catalog No. AAB242120B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4130-08-9
    Molecular Formula C8H12O6Si
    Formula Weight 232.26
    MDL Number MFCD00014975
    UN Number UN1760
    Beilstein 1792454
    Synonym Vinyltriacetoxysilane
    Chemical Name or Material Triacetoxy(vinyl)silane

    Specifications

    Formula Weight 232.26
    TSCA TSCA
    Molecular Formula C8H12O6Si
    MDL Number MFCD00014975
    UN Number UN1760
    Synonym Vinyltriacetoxysilane
    Boiling Point 112° to 113°C (1mmHg)
    Quantity 1000g
    Flash Point 76°C (168°F)
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    CAS 4130-08-9
    Beilstein 1792454
    Melting Point 7°C
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
    Chemical Name or Material Triacetoxy(vinyl)silane
    Assay 95%

    Description

    Applications
    Triacetoxy(vinyl)silane is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit