Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triallyl 1,3,5-Benzenetricarboxylate 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
17832-16-5
C18H18O6
330.336
MFCD00008647
VOSUIKFOFHZNED-UHFFFAOYSA-N
triallyl trimesate,triallyl benzene-1,3,5-tricarboxylate,triallyl 1,3,5-benzenetricarboxylate,triallylbenzene-1,3,5-tricarboxylate,1,3,5-benzenetricarboxylic acid, tri-2-propenyl ester,1,3,5-benzenetricarboxylic acid, 1,3,5-tri-2-propen-1-yl ester,1,3,5-benzenetricarboxylic acid triallyl ester,1,3,5-tris prop-2-en-1-yl benzene-1,3,5-tricarboxylate,triallyltrimesate,acmc-1c8hf
87328
tris(prop-2-enyl) benzene-1,3,5-tricarboxylate
C=CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC=C)C(=O)OCC=C
17832-16-5
C18H18O6
330.336
MFCD00008647
VOSUIKFOFHZNED-UHFFFAOYSA-N
triallyl trimesate,triallyl benzene-1,3,5-tricarboxylate,triallyl 1,3,5-benzenetricarboxylate,triallylbenzene-1,3,5-tricarboxylate,1,3,5-benzenetricarboxylic acid, tri-2-propenyl ester,1,3,5-benzenetricarboxylic acid, 1,3,5-tri-2-propen-1-yl ester,1,3,5-benzenetricarboxylic acid triallyl ester,1,3,5-tris prop-2-en-1-yl benzene-1,3,5-tricarboxylate,triallyltrimesate,acmc-1c8hf
87328
tris(prop-2-enyl) benzene-1,3,5-tricarboxylate
C=CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC=C)C(=O)OCC=C
Quantity 25g
Color White-Yellow
Boiling Point 212°C
Formula Weight 330.34
Physical Form Crystal-Lump at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Triallyl 1,3,5-Benzenetricarboxylate