Alfa Aesar™ Triallyl phosphite

Manufacturer: Alfa Aesar™ 1748430

Quantity 250g

 View more versions of this product

Catalog No. AA1748430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 102-84-1
    Linear Formula (CH2=CHCH2O)3P
    Formula Weight 202.19
    MDL Number MFCD00014948
    UN Number UN3278
    Beilstein 1705535
    Chemical Name or Material Triallyl phosphite

    Specifications

    Formula Weight 202.19
    Health Hazard 3 GHS P Statement
    P280h-P309-P310
    Wear protective gloves/clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Physical Form Liquid
    MDL Number MFCD00014948
    Boiling Point 84°C (12mmHg)
    Flash Point 86° (187°F)
    Odor Strong
    Sensitivity Air & Moisture Sensitive
    TSCA TSCA
    CAS 102-84-1
    Beilstein 1705535
    Color Colorless
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
    Linear Formula (CH2=CHCH2O)3P
    UN Number UN3278
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H301-H312
    Toxic if swallowed.
    Harmful in contact with skin.
    RTECS Number TH1772500
    Chemical Name or Material Triallyl phosphite

    GHS H Statement
    H301-H312
    Toxic if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280h-P309-P310
    Wear protective gloves/clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit