Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triallyl phosphite

Manufacturer:  Alfa Aesar™ 1748409

Quantity 10g

 View more versions of this product

Catalog No. AA1748409


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

102-84-1
202.19
UN3278
Triallyl phosphite
(CH2=CHCH2O)3P
MFCD00014948
1705535

Specifications

102-84-1
(CH2=CHCH2O)3P
MFCD00014948
1705535
84°C (12mmHg)
86° (187°F)
Air & Moisture Sensitive
Liquid
202.19
UN3278
Colorless
10g
Strong
Triallyl phosphite

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.

RTECSNumber : TH1772500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit