Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%, Stabilized, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
1025-15-6
C12H15N3O3
249.27
MFCD00006554
KOMNUTZXSVSERR-UHFFFAOYSA-N
triallyl isocyanurate,isocyanuric acid triallyl ester,1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione,triallylisocyanurate,taic,diak 7,1,3,5-triallylisocyanurate,1,3,5-triallyl isocyanurate,1,3,5-triallylisocyanuric acid,1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
13931
1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C
1025-15-6
C12H15N3O3
249.27
MFCD00006554
KOMNUTZXSVSERR-UHFFFAOYSA-N
triallyl isocyanurate,isocyanuric acid triallyl ester,1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione,triallylisocyanurate,taic,diak 7,1,3,5-triallylisocyanurate,1,3,5-triallyl isocyanurate,1,3,5-triallylisocyanuric acid,1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
13931
1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C
Quantity 5g
Density 1.159g/mL
Infrared Spectrum Authentic
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Stabilizer 1200ppm MEHQ
Color Colorless to Yellow
Assay Percent Range 98%
Beilstein 26, IV, 637
Formula Weight 249.27
Physical Form Liquid After Melting
Percent Purity ≥97.5%
Chemical Name or Material Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 213-834-7

RTECSNumber : XZ1915000

TSCA : TSCA