Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%, Stabilized, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
1025-15-6
C12H15N3O3
249.27
MFCD00006554
KOMNUTZXSVSERR-UHFFFAOYSA-N
triallyl isocyanurate,isocyanuric acid triallyl ester,1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione,triallylisocyanurate,taic,diak 7,1,3,5-triallylisocyanurate,1,3,5-triallyl isocyanurate,1,3,5-triallylisocyanuric acid,1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
13931
1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C
1025-15-6
C12H15N3O3
249.27
MFCD00006554
KOMNUTZXSVSERR-UHFFFAOYSA-N
triallyl isocyanurate,isocyanuric acid triallyl ester,1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione,triallylisocyanurate,taic,diak 7,1,3,5-triallylisocyanurate,1,3,5-triallyl isocyanurate,1,3,5-triallylisocyanuric acid,1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
13931
1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C
Boiling Point 149.0°C to 152.0°C (4.0mmHg)
Specific Gravity 1.159
Packaging Glass bottle
Stabilizer 1200ppm MEHQ
Refractive Index 1.5120 to 1.5140
Viscosity 90.1 mPa.s (30°C)
Infrared Spectrum Authentic
Quantity 500g
Color Colorless to Yellow
Density 1.1590g/mL
Melting Point 24.8°C
Flash Point 245°C
Assay Percent Range 97.5% min. (GC)
Beilstein 26, IV, 637
Solubility Information Solubility in water: >1g/L (20°C). Other solubilities: soluble in acetone,dimethyl sulfoxide,ethanol
Formula Weight 249.27
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Harmful in contact with skin.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

EINECSNumber : 213-834-7

RTECSNumber : XZ1915000

TSCA : TSCA