Alfa Aesar™ Triallylamine, 98%

Manufacturer: Alfa Aesar™ A1208118

Quantity 50g

 View more versions of this product

Catalog No. AAA1208118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 102-70-5
    Molecular Formula (H2C=CHCH2)3N
    Formula Weight 137.23
    MDL Number MFCD00026093
    UN Number UN2610
    Chemical Name or Material Triallylamine

    Specifications

    Formula Weight 137.23
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 102-70-5
    Melting Point −70°C
    Boiling Point 150° to 151°C
    Health Hazard 1 Danger
    Flash Point 31°C (87°F)
    Odor Amine-like
    Health Hazard 2 GHS H Statement
    H311-H314-H318-H226-H302-H332
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.
    Molecular Formula (H2C=CHCH2)3N
    MDL Number MFCD00026093
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIALLYLAMINE
    RTECS Number XX5950000
    UN Number UN2610
    Quantity 50g
    Chemical Name or Material Triallylamine
    Assay 98%

    Description

    Applications
    It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Store away from strong oxidizing agents and acids. Protect against electrostatic charges and keep away from ignition sources.

    GHS H Statement
    H311-H314-H318-H226-H302-H332
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit