Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamcinolone Acetonide, Powder, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
76-25-5
C24H31FO6
434.50
YNDXUCZADRHECN-JNQJZLCISA-N
(1S,2S,4R,8S,9S,11S,12R,13S)-12-fluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
76-25-5
C24H31FO6
434.50
YNDXUCZADRHECN-JNQJZLCISA-N
(1S,2S,4R,8S,9S,11S,12R,13S)-12-fluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

TR107, 76-25-5, C24H31FO6

Spectrum™ Triamcinolone Acetonide, Powder, USP is a topical corticosteroid used to treat a variety of skin conditions (e.g., eczema, dermatitis, allergies, rash). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Bottle
Loss on Drying 0.015
Specific Rotation +118° to +130°
Quantity 100 g
CAS Max % 1
Percent Purity 97 to 102%
Grade USP