Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamcinolone Diacetate, Micronized, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
67-78-7
C25H31FO8
478.51
XGMPVBXKDAHORN-RBWIMXSLSA-N
2-[(1S,2R,3aS,3bS,9aS,9bR,10S,11aS)-2-(acetyloxy)-9b-fluoro-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
CC(=O)OCC(=O)[C@@]1(O)[C@@H](C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)OC(C)=O
67-78-7
C25H31FO8
478.51
XGMPVBXKDAHORN-RBWIMXSLSA-N
2-[(1S,2R,3aS,3bS,9aS,9bR,10S,11aS)-2-(acetyloxy)-9b-fluoro-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
CC(=O)OCC(=O)[C@@]1(O)[C@@H](C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)OC(C)=O

TR109, 67-78-7, C25H31FO8

Spectrum™ Triamcinolone Diacetate, Micronized, USP is a topical corticosteroid used to treat a variety of skin conditions (e.g., eczema, dermatitis, allergies, rash). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.005
Water 0.06
Specific Rotation +39° to +45°
Quantity 25 g
CAS Max % 1
Percent Purity 97 to 103%
Grade USP