Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamcinolone Hexacetonide, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
5611-51-8
C30H41FO7
532.65
TZIZWYVVGLXXFV-FLRHRWPCSA-N
2-[(1S,2S,4R,8S,9S,11S,12R,13S)-12-fluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-8-yl]-2-oxoethyl 3,3-dimethylbutanoate
CC(C)(C)CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C
5611-51-8
C30H41FO7
532.65
TZIZWYVVGLXXFV-FLRHRWPCSA-N
2-[(1S,2S,4R,8S,9S,11S,12R,13S)-12-fluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-8-yl]-2-oxoethyl 3,3-dimethylbutanoate
CC(C)(C)CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C

T2049, 5611-51-8, C30H41FO7

Spectrum™ Triamcinolone Hexacetonide, USP is a topical corticosteroid used to treat a variety of skin conditions (e.g., eczema, dermatitis, allergies, rash). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.02
Optical Rotation +91° to +98°
Quantity 25 g
CAS Max % 1
Percent Purity 97 to 102%