Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamcinolone, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
124-94-7
C21H27FO6
394.44
GFNANZIMVAIWHM-OBYCQNJPSA-N
(1S,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,2,10-trihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
C[C@]12C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)[C@]2(O)C(=O)CO
124-94-7
C21H27FO6
394.44
GFNANZIMVAIWHM-OBYCQNJPSA-N
(1S,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,2,10-trihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
C[C@]12C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)[C@]2(O)C(=O)CO

TR103, 124-94-7, C21H27FO6

Spectrum™ Triamcinolone, USP is a topical corticosteroid used to treat a variety of skin conditions (e.g., eczema, dermatitis, allergies, rash). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Quantity 250 mg
Loss on Drying 0.02
Specific Rotation +65° to +72°
Ignition Residue 0.005
CAS Max % 1
Percent Purity 97 to 102%
Grade USP