Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamterene 98.0+%, TCI America™

Click to view available options
Quantity:
25g
396-01-0
C12H11N7
253.269
MFCD00006708
FNYLWPVRPXGIIP-UHFFFAOYSA-N
triamterene,2,4,7-triamino-6-phenylpteridine,dyrenium,triamteren,dytac,pterofen,pterophene,triamteril,triteren,ademin
5546
6-phenylpteridine-2,4,7-triamine
C1=CC=C(C=C1)C2=NC3=C(N=C2N)N=C(N=C3N)N
396-01-0
C12H11N7
253.269
MFCD00006708
FNYLWPVRPXGIIP-UHFFFAOYSA-N
triamterene,2,4,7-triamino-6-phenylpteridine,dyrenium,triamteren,dytac,pterofen,pterophene,triamteril,triteren,ademin
5546
6-phenylpteridine-2,4,7-triamine
C1=CC=C(C=C1)C2=NC3=C(N=C2N)N=C(N=C3N)N
Quantity 25g
Color Yellow
Formula Weight 253.27
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Triamterene

RTECSNumber : UO3470000

TSCA : No