General Data Healthcare™ General Data Healthcare Systems™ RTP™/RTPH-300 Rapid Tissue Processor

Directs flow of all reagents, waxes and cleaning solutions to and from the processing chamber through a single rotary valve within a heated chamber for increased reliability

Manufacturer: General Data Healthcare™ RTPH300

 View more versions of this product

Catalog No. 22-046-857

  Check Availability
Add to Cart

    Specifications

    Product Type RTPH-300 Rapid Tissue Processor w/RHE Rapid Heat Exchange
    Height (Metric) Module 1300mm
    Weight (English) Module 396.83 lb.
    Weight (Metric) Module 180kg
    Voltage 100 to 240V

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit