Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIB1 Mouse anti-Human, Clone: OTI8C8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526721

Catalog No. PIMA526721


Add to cart

Description

Description

TRIB1 antibody, also called gprotein coupled receptor induced, skip1, gig2, c8fwATP, functions in binding protein binding, protein kinase inhibitor activity, and protein serine/threonine kinase activities. Overexpression of TRIB1 in HeLa cells repressed the basal activity of the IL8 promoter by inhibiting AP1 activity. Overexpression of TRIB1 inhibited oncogenic Ras -driven AP1 activation and MEKK1-mediated AP1 activation. ERK activation was enhanced by TRIB1. Coimmunoprecipitation and yeast 2-hybrid assays showed that MEK1 interacted with both TRIB1 and TRIB3, and MKK4 interacted specifically with TRIB1. Cotransfection of MKK4 enhanced the level of TRIB1, indicating that the TRIB-MAPKK interaction stabilized TRIB1.
Specifications

Specifications

TRIB1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96RU8
C8FW, GIG2, SKIP1, TRB1, G-protein-coupled receptor induced protein, G-protein-coupled receptor-induced gene 2 protein, G-protein-coupled receptor-induced protein 2, GIG-2, TRB-1, phosphoprotein regulated by mitogenic pathways, tribbles homolog 1, tribbles-like protein 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10221
Immunohistochemistry (Paraffin), Western Blot
OTI8C8
Unconjugated
TRIB1
Liquid
TRIB1
Full length human recombit protein of TRIB1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit