Learn More
TRIB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153591AP
Catalog No. 50-173-0176
Add to cart
Description
Description
This gene encodes one of three members of the Tribbles family. The Tribbles members share a Trb domain, which is homologous to protein serine-threonine kinases, but lacks the active site lysine and probably lacks a catalytic function. The Tribbles proteins interact and modulate the activity of signal transduction pathways in a number of physiological and pathological processes. This Tribbles member induces apoptosis of cells mainly of the hematopoietic origin. It has been identified as a protein up-regulated by inflammatory stimuli in myeloid (THP-1) cells, and also as an oncogene that inactivates the transcription factor C/EBPalpha (CCAAT/enhancer-binding protein alpha) and causes acute myelogenous leukemia. Alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
TRIB2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K4K3, Q92519
C5FW, GS3955, TRB 2, TRB2, TRIB2, Tribbles homolog 2
Rabbit
IgG
150 μL
-20°C
Primary
217410, 28951, 313974
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TRIB2
Liquid
TRIB2
TRIB2 Fusion Protein Ag7605
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat