Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tribasic Calcium Phosphate, Powder, NF, 34-40%, Spectrum™

Click to view available options
Quantity:
100 lbs.
12 kg
2.5 kg
Packaging:
Fiber Drum
Poly Pail
12167-74-7
Ca3O8P2
310.17
QORWJWZARLRLPR-UHFFFAOYSA-H
tricalcium diphosphate
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
12167-74-7
Ca3O8P2
310.17
QORWJWZARLRLPR-UHFFFAOYSA-H
tricalcium diphosphate
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

CA165, 12167-74-7, Ca5(OH)(PO4)3

Spectrum™ Tribasic Calcium Phosphate, Powder, NF is a widely used pharmaceutical excipient as binder and filler in solid oral dosage forms which include compressed tablets and hard gelatin capsules. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Arsenic (As) 3 ppm
Ignition Residue 0.08
Percent Purity 34 to 40%
Grade NF
Packaging Fiber Drum
Quantity 12 kg