Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tribenzylamine, 99+%

Click to view available options
Quantity:
25g
100g
620-40-6
C21H21N
287.406
MFCD00004773
MXHTZQSKTCCMFG-UHFFFAOYSA-N
tribenzylamine,benzenemethanamine, n,n-bis phenylmethyl,unii-hz10o1931j,trisbenzylamine,n,n-dibenzyl-1-phenyl-methanamine,pubchem21076,acmc-1atok,dsstox_cid_27031,dsstox_rid_82052
24321
N,N-dibenzyl-1-phenylmethanamine
C1=CC=C(C=C1)CN(CC2=CC=CC=C2)CC3=CC=CC=C3
620-40-6
C21H21N
287.406
MFCD00004773
MXHTZQSKTCCMFG-UHFFFAOYSA-N
tribenzylamine,benzenemethanamine, n,n-bis phenylmethyl,unii-hz10o1931j,trisbenzylamine,n,n-dibenzyl-1-phenyl-methanamine,pubchem21076,acmc-1atok,dsstox_cid_27031,dsstox_rid_82052
24321
N,N-dibenzyl-1-phenylmethanamine
C1=CC=C(C=C1)CN(CC2=CC=CC=C2)CC3=CC=CC=C3
Flash Point 203°C (397°F)
Melting Point 91°C to 94°C
Odor Weak
Boiling Point 230°C (13mmHg)
Quantity 25g
Beilstein 2214682
Formula Weight 287.41
Percent Purity ≥99%
Chemical Name or Material Tribenzylamine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA