Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tribromoacetic acid, 97%

Click to view available options
Quantity:
5g
25g
75-96-7
C2HBr3O2
296.74
MFCD00004167
QIONYIKHPASLHO-UHFFFAOYSA-N
tribromoacetic acid,acetic acid, tribromo,tribromacetic acid,unii-71ktl99qjx,ccris 7887,71ktl99qjx,acetic acid, 2,2,2-tribromo,snpljqbkdvidtaetp@,dsstox_cid_1668,acmc-209p0y
6415
2,2,2-tribromoacetic acid
C(=O)(C(Br)(Br)Br)O
75-96-7
C2HBr3O2
296.74
MFCD00004167
QIONYIKHPASLHO-UHFFFAOYSA-N
tribromoacetic acid,acetic acid, tribromo,tribromacetic acid,unii-71ktl99qjx,ccris 7887,71ktl99qjx,acetic acid, 2,2,2-tribromo,snpljqbkdvidtaetp@,dsstox_cid_1668,acmc-209p0y
6415
2,2,2-tribromoacetic acid
C(=O)(C(Br)(Br)Br)O
Melting Point 130°C to 133°C
Quantity 25g
Boiling Point 244°C to 246°C
UN Number UN3261
Beilstein 1755150
Merck Index 14,9609
Formula Weight 296.74
Percent Purity 97%
Chemical Name or Material Tribromoacetic acid

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA