Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tribromoacetic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
75-96-7
C2HBr3O2
296.73
MFCD00004167
QIONYIKHPASLHO-UHFFFAOYSA-N
tribromoacetic acid,acetic acid, tribromo,tribromacetic acid,unii-71ktl99qjx,ccris 7887,71ktl99qjx,acetic acid, 2,2,2-tribromo,snpljqbkdvidtaetp@,dsstox_cid_1668,acmc-209p0y
6415
2,2,2-tribromoacetic acid
C(=O)(C(Br)(Br)Br)O
75-96-7
C2HBr3O2
296.73
MFCD00004167
QIONYIKHPASLHO-UHFFFAOYSA-N
tribromoacetic acid,acetic acid, tribromo,tribromacetic acid,unii-71ktl99qjx,ccris 7887,71ktl99qjx,acetic acid, 2,2,2-tribromo,snpljqbkdvidtaetp@,dsstox_cid_1668,acmc-209p0y
6415
2,2,2-tribromoacetic acid
C(=O)(C(Br)(Br)Br)O
Boiling Point 245°C
Color White to Yellow
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Melting Point 130°C to 133°C
Assay Percent Range 99%
Linear Formula CBr3CO2H
Beilstein 02, 220
Merck Index 15, 9770
Formula Weight 296.73
Physical Form Crystals
Percent Purity ≥98.5%
Chemical Name or Material Tribromoacetic acid, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-919-9

TSCA : TSCA