Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl(1-ethoxyvinyl)tin, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
97674-02-7
C16H34OSn
361.16
MFCD00010240
HGXJOXHYPGNVNK-UHFFFAOYSA-N
tributyl 1-ethoxyvinyl stannane,tributyl 1-ethoxyvinyl tin,tributyl 1-ethoxyethenyl stannane,1-ethoxyvinyltributyltin,1-ethoxyvinyl tributyltin,1-ethoxyvinyltri-n-butyltin,1-ethoxyvinyl tributylstannane,1-ethoxyethenyl tributylstannane,tributyl-1-ethoxyvinyl tin,stannane, tributyl 1-ethoxyethenyl
619414
tributyl(1-ethoxyethenyl)stannane
CCCC[Sn](CCCC)(CCCC)C(=C)OCC
97674-02-7
C16H34OSn
361.16
MFCD00010240
HGXJOXHYPGNVNK-UHFFFAOYSA-N
tributyl 1-ethoxyvinyl stannane,tributyl 1-ethoxyvinyl tin,tributyl 1-ethoxyethenyl stannane,1-ethoxyvinyltributyltin,1-ethoxyvinyl tributyltin,1-ethoxyvinyltri-n-butyltin,1-ethoxyvinyl tributylstannane,1-ethoxyethenyl tributylstannane,tributyl-1-ethoxyvinyl tin,stannane, tributyl 1-ethoxyethenyl
619414
tributyl(1-ethoxyethenyl)stannane
CCCC[Sn](CCCC)(CCCC)C(=C)OCC
Packaging Glass bottle
Specific Gravity 1.069
Flash Point 113°C
Refractive Index 1.4750 to 1.4770
Color Colorless
Density 1.0690g/mL
Quantity 1g
Boiling Point 85.0°C to 86.0°C (0.1mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Linear Formula [CH3(CH2)3]SnC(CH2)OCH2CH3
Solubility Information Solubility in water: insoluble
Formula Weight 361.16
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Tributyl(1-ethoxyvinyl)tin
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Toxic if swallowed.
Harmful

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN

GHS Signal Word: Danger