Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl(3-methyl-2-butenyl)tin, 90%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
53911-92-5
C17H36Sn
359.185
MFCD01863649
XEFRYQPTIWSVGI-UHFFFAOYSA-N
tributyl 3-methylbut-2-en-1-yl stannane,tributyl 3-methyl-2-butenyl tin,tributylprenylstannane,acmc-1am9p,tri-n-butyl 3-methyl-2-butenyl tin,tributyl 3-methylbut-2-enyl stannane,stannane, tributyl 3-methyl-2-butenyl,tributyl 3-methylbut-2-en-1-yl ;stannane
2734722
tributyl(3-methylbut-2-enyl)stannane
CCCC[Sn](CCCC)(CCCC)CC=C(C)C
53911-92-5
C17H36Sn
359.185
MFCD01863649
XEFRYQPTIWSVGI-UHFFFAOYSA-N
tributyl 3-methylbut-2-en-1-yl stannane,tributyl 3-methyl-2-butenyl tin,tributylprenylstannane,acmc-1am9p,tri-n-butyl 3-methyl-2-butenyl tin,tributyl 3-methylbut-2-enyl stannane,stannane, tributyl 3-methyl-2-butenyl,tributyl 3-methylbut-2-en-1-yl ;stannane
2734722
tributyl(3-methylbut-2-enyl)stannane
CCCC[Sn](CCCC)(CCCC)CC=C(C)C
Refractive Index 1.4860 to 1.4900 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color Colorless
Density 1.0690g/mL
Assay Percent Range 90%
Linear Formula [CH3(CH2)3]3SnCH2CH=C(CH3)2
Formula Weight 359.18
Physical Form Liquid
Percent Purity ≥88%
Chemical Name or Material Tributyl(3-methyl-2-butenyl)tin, 90%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Toxic if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger