Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl(4-fluorophenyl)stannane, 95%, Maybridge

Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
17151-47-2
C18H31FSn
385.154
MFCD01313817
FLPKMHDTSOPPOJ-UHFFFAOYSA-N
tributyl 4-fluorophenyl stannane,tributyl-4-fluorophenyl stannane,4-fluoro-tri-n-butylstannyl benzene,4-fluorophenyltributyltin,4-fluorophenyl tributylstannane,4-fluorophenyl-tri-n-butylstannane,4-tri-n-butylstannyl fluorobenzene,4-fluoro-tributylstannyl-benzene,stannane, tributyl 4-fluorophenyl,tri-n-butyl-4-fluorophenyl stannane
2737524
tributyl-(4-fluorophenyl)stannane
CCCC[Sn](CCCC)(CCCC)C1=CC=C(C=C1)F
17151-47-2
C18H31FSn
385.154
MFCD01313817
FLPKMHDTSOPPOJ-UHFFFAOYSA-N
tributyl 4-fluorophenyl stannane,tributyl-4-fluorophenyl stannane,4-fluoro-tri-n-butylstannyl benzene,4-fluorophenyltributyltin,4-fluorophenyl tributylstannane,4-fluorophenyl-tri-n-butylstannane,4-tri-n-butylstannyl fluorobenzene,4-fluoro-tributylstannyl-benzene,stannane, tributyl 4-fluorophenyl,tri-n-butyl-4-fluorophenyl stannane
2737524
tributyl-(4-fluorophenyl)stannane
CCCC[Sn](CCCC)(CCCC)C1=CC=C(C=C1)F
Packaging Amber Glass Bottle
Quantity 1g
CAS Min % 95.0
Assay Percent Range 95%
Formula Weight 385.15
Percent Purity 95%
Chemical Name or Material Tributyl(4-fluorophenyl)stannane

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
Harmful in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger